Dimethyl-3-oxoglutarate 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate
115700250 115701000 115705000
ACRO115700250EA 685 SEK
ACRO115700250 ACRO115701000 ACRO115705000
Dimethyl-3-oxoglutarate 97%
Dimethyl-3-oxoglutarate
Formel: C₇H₁₀O₅
MW: 174.15 g/mol
Kokpunkt: 237…241 °C (760 mmHg)
Smältpunkt: 16…17 °C
Densitet: 1.18 g/cm³
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00008462
CAS nummer: 1830-54-2
EINECS: 217-385-8
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20°C, 589 nm)
GC ≥96.0 %
Visual transition interval pH 9.0 (blue)