Diethyl acetamidomalonate 99+%
Leverantör: Thermo Fisher Scientific
Varning
113931000 113935000 113930250
ACRO113931000EA 825 SEK
ACRO113931000 ACRO113935000 ACRO113930250
Diethyl acetamidomalonate 99+%
Diethyl acetamidomalonate
Formel: C₉H₁₅NO₅
Kokpunkt: 185 °C (20 mmHg)
Smältpunkt: 94…97 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009146
CAS nummer: 1068-90-2
EINECS: 213-952-9
Order Now
Specifikation testresultat
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 97 °C
GC ≥99.0 %
BSC Certification Passes