Dibenzyl phosphonate 90+%, Technical Grade
Leverantör: Thermo Scientific
Varning
Synonyms: Dibenzyl phosphite
112660250 112660050 112661000
ACRO112660250EA 1375 SEK
ACRO112660250 ACRO112660050 ACRO112661000
Dibenzyl phosphonate 90+%, Technical Grade
Dibenzyl phosphonate
Formel: C₁₄H₁₅O₃P
MW: 262,25 g/mol
Kokpunkt: 110…120 °C (,1 mmHg)
Densitet: 1,18 g/cm³
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00004774
CAS nummer: 17176-77-1
EINECS: 241-226-1
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5520 to 1.5550 (20°C, 589 nm)
NMR ≥90.0 %
UV at 280 nm A: ≤0.05
pH 5.5 to 8.0 (5 % in water)
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Heavy metals (as Pb) ≤5 ppm