Diallylamine 97%
Leverantör: Thermo Fisher Scientific
Fara
112040010 112042500
ACRO112040010EA 2675 SEK
ACRO112040010 ACRO112042500
Diallylamine 97%
Diallylamine
Formel: C₆H₁₁N
Smältpunkt: –88 °C
Densitet: 0,789 g/cm³
Flampunkt: 7 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008642
CAS nummer: 124-02-7
EINECS: 204-671-2
UN: 2359
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4395 to 1.4415 (20°C, 589 nm)
GC ≥96.0 %
UV at 450 nm A: ≤0.01
Color scale ≤20 APHA