Bariumnitrat ≥99.95%
Leverantör: Thermo Fisher Scientific
Varning
30481.14 30481.22
30481.14EA 1175 SEK
30481.14 30481.22
Bariumnitrat ≥99.95%
Barium nitrate
Purity: 99.95% CAS No.: 10022-31-8 Molecular Formula: Ba(NO3)2 Formula Weight: 261.35 Physical Form: Crystalline Density: 3.24 Melting Point: 592° dec. Merck Reference: 14,983 UN No.: 1446 MDL No.: MFCD00003442
Formel: Ba(NO₃)₂
MW: 261.34 g/mol
Smältpunkt: 592 °C (dec.)
Densitet: 3.24
Storage Temperature: Omgivande
MDL Number: MFCD00003442
CAS nummer: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1 II
Merck Index: 14,983
Order Now
SPECIFIKATIONER
Environmentally Preferable