Magnesiumklorid, ultra dry ≥99.9%
Leverantör: Thermo Fisher Scientific
Varning
Ampuled under argon
42843.06 42843.14 42843.22
42843.06EA 1500 SEK
42843.06 42843.14 42843.22
Magnesiumklorid, ultra dry ≥99.9%
Magnesiumklorid
Purity: 99.9% CAS No.: 7786-30-3 Molecular Formula: Cl2Mg Formula Weight: 95.22 Physical Form: Powder, Ampouled under argon Density: 2.32 Appearance: White to yellow Odour: Odourless Melting Point: 714° Merck Reference: 14,5662 MDL No.: MFCD00011106
Formel: MgCl₂
MW: 95.21 g/mol
Kokpunkt: 1412
Smältpunkt: 714 °C
Densitet: 2.32
Storage Temperature: Omgivande
MDL Number: MFCD00011106
CAS nummer: 7786-30-3
EINECS: 232-094-6
Merck Index: 14,5662