Magnesiumklorid, vattenfri 99%
Leverantör: Thermo Fisher Scientific
Varning
12315.A1 12315.A7 12315.C4
12315.A1EA 1450 SEK
12315.A1 12315.A7 12315.C4
Magnesiumklorid, vattenfri 99%
Magnesiumklorid
Purity: 99% CAS No.: 7786-30-3 Molecular Formula: MgCl2 Formula Weight: 95.22 Physical Form: Crystalline Density: 2.32 Appearance: White to yellow Odour: Odourless Melting Point: 714° Solubility: Soluble in water with evolution of heat Merck Reference: 14,5662 MDL No.: MFCD00011106
Catalyst, flocculating agent, various magnesium cements, ceramics, analytical reagent
Formel: MgCl₂
MW: 95.21 g/mol
Kokpunkt: 1412
Smältpunkt: 714 °C
Densitet: 2.32
Storage Temperature: Omgivande
MDL Number: MFCD00011106
CAS nummer: 7786-30-3
EINECS: 232-094-6
Merck Index: 14,5662