Natriummetasilikat, vattenfri, granular, tech. –18 Mesh
Leverantör: Thermo Scientific
Fara
Synonyms: Sodium metasilicate
14106.A1 14106.D9 14106.99
14106.A1EA 1500 SEK
14106.A1 14106.D9 14106.99
Natriummetasilikat, vattenfri, granular, tech. –18 Mesh
Natriummetasilikat
Sodium metasilicate is used as a starting material for zeolites and silica catalysts. It acts as an adhesive and binder, corrosion inhibitor, penetrating sealant, coagulant in waste water treatment, fixative for hand dyeing with reactive dye and detergent auxiliaries.
It is also used in boiler compounds, cleaners, bleaches, aluminum paint strippers and washing carbonated drinking bottles. It finds application in cements, drilling fluids, fireproofing wood, refractories and automobiles. Further, it serves as a surfactant, emulsifier, wetting agent, suspending agent, dispersing agent and anti-corrosive agent in boiler water.
Incompatible with acids, bronze, aluminum, brass, copper, lead, tin, zinc and other alkali sensitive metals.
Formel: Na₂O₃Si
MW: 122,06 g/mol
Smältpunkt: 1088 °C
Densitet: 2,61
Storage Temperature: Omgivande
MDL Number: MFCD00003492
CAS nummer: 6834-92-0
EINECS: 229-912-9
UN: 3253
ADR: 8,III
Merck Index: 14,8642
Order Now
Specifikation testresultat
Assay (Unspecified), SiO₂ 44.7 - 47.6%
Assay (Unspecified), Na₂O 49.1 - 51.7%