Natriumvätesulfat 92% tech., granular
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Natriumbisulfat
214850010 214850250 214850025
ACRO214850010EA 920 SEK
ACRO214850010 ACRO214850250 ACRO214850025
Natriumvätesulfat 92% tech., granular
Natriumvätesulfat
Formel: HNaO₄S
MW: 120.06 g/mol
Smältpunkt: 177…180 °C
Densitet: 2.1 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00003531
CAS nummer: 7681-38-1
EINECS: 231-665-7
Merck Index: 12,08729
Order Now
Specifikation testresultat
Appearance White to light yellow Granules
Titration with NaOH ≥90 % (≥37.6 % (equivalent H2SO4))
Chloride (Cl) ≤1000 ppm
Iron (Fe) ≤200 ppm