Natriumsulfat, anhydrous 99%
Leverantör: Thermo Fisher Scientific
Synonyms: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt
A19890.0B A19890.0I A19890.36
A19890.0BEA 750 SEK
A19890.0B A19890.0I A19890.36
Natriumsulfat, anhydrous 99%
Sodium sulphate
Purity: 99% CAS No.: 7757-82-6 Molecular Formula: Na2O4S Formula Weight: 142.04 Refractive Index: 1.484 Odour: Odourless Merck Reference: 14,8680 MDL No.: MFCD00003504
Formel: Na₂SO₄
MW: 142.04 g/mol
Kokpunkt: 1429 °C (1013 hPa)
Smältpunkt: 884 °C
Densitet: 2.680
Storage Temperature: Omgivande
MDL Number: MFCD00003504
CAS nummer: 7757-82-6
EINECS: 231-820-9
Merck Index: 14,8680
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM