Litiumsulfat, vattenfri ≥99.7%
Leverantör: Thermo Scientific
Varning
13404.30 13404.A1 13404.A7
13404.30EA 1700 SEK
13404.30 13404.A1 13404.A7
Litiumsulfat, vattenfri ≥99.7%
Litiumsulfat
Purity: 99.7% CAS No.: 10377-48-7 Molecular Formula: Li2SO4 Formula Weight: 109.94 Physical Form: -40 Mesh Powder Density: 2.22 Appearance: White Odour: Odourless Melting Point: 845° Merck Reference: 14,5541 MDL No.: MFCD00011086
Formel: Li₂O₄S
MW: 109,95 g/mol
Kokpunkt: 1377 °C (1013 hPa)
Smältpunkt: 845 °C
Densitet: 2,22
Storage Temperature: Omgivande
MDL Number: MFCD00011086
CAS nummer: 10377-48-7
EINECS: 233-820-4
Merck Index: 14,5541