Natriumtiocyanat, Supelco®
Leverantör: Merck
Varning
Synonyms: Sodium rhodanide
M-Clarity™ quality level = MQ200
1.06627.2500 1.06627.9025
1.06627.2500EA 2220 SEK
1.06627.2500 1.06627.9025
Natriumtiocyanat, Supelco®
Natriumtiocyanat
Formel: NaSCN
MW: 81.07 g/mol
Smältpunkt: 287
Densitet: 1.735 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011123
CAS nummer: 540-72-7
EINECS: 208-754-4
Merck Index: 13,09401
Order Now
Specifikation testresultat
Solubility 1200 g/l (20 °C)
Melting Point 310 °C
Molar Mass 81.07 g/mol
Bulk Density 800 - 1000 kg/m3
Vapor Pressure  - 1 hPa (20 °C)
Density 1.74 g/cm3 (20 °C)
pH 6 - 8 (100 g/l, H2O, 20 °C)
Synonyms Sodium sulfocyanide
Hill Formula CNNaS
Chemical Formula NaSCN