Natriumperoxodisulfat 98+%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Sodium peroxidisulphate, Sodium persulphate, Disodium peroxodisulphate, Peroxydisulphuric acid disodium salt
202020010 202020050 202025000
ACRO202020010EA 1425 SEK
ACRO202020010 ACRO202020050 ACRO202025000
Natriumperoxodisulfat 98+%
Sodium peroxodisulphate
Formel: Na₂S₂O₈
Smältpunkt: 100 °C
Densitet: 2,4 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003501
CAS nummer: 7775-27-1
EINECS: 231-892-1
UN: 1505
ADR: 5.1,III
Merck Index: 13,08729
Order Now
Specifikation testresultat
Appearance White to almost white Powder or crystals
Titration Iodometric 98.0 to 102.0 %
Water ≤0.15 %
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤20 ppm
Total nitrogen ≤0.2 %