Natriumditionit ≥85%, tech.
Leverantör: Thermo Scientific
Fara
Synonyms: Sodium hydrosulphite
33381.A1 33381.14 33381.22
33381.A1EA 2000 SEK
33381.A1 33381.14 33381.22
Natriumditionit ≥85%, tech.
Natriumditionit
Purity: 85+% CAS No.: 7775-14-6 Molecular Formula: Na2S2O4 Formula Weight: 174.11 Physical Form: Powder Sensitivity: Moisture Sensitive Merck Reference: 14,8626 UN No.: 1384 MDL No.: MFCD00011640
Formel: Na₂S₂O₄
MW: 174,11 g/mol
Smältpunkt: 52 °C (dec.)
Densitet: 2,19 g/cm³ (20 °C)
Flampunkt: 101 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011640
CAS nummer: 7775-14-6
EINECS: 231-890-0
UN: 1384
ADR: 4.2,II
Merck Index: 14,8626