trinatriumortofosfat, vattenfri, tech.
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: tri-Sodium phosphate, Sodium phosphate tribasic, Trisodium phosphate
13438.A1 13438.A7
13438.A1EA 1300 SEK
13438.A1 13438.A7
trinatriumortofosfat, vattenfri, tech.
trinatriumortofosfat
CAS No.: 7601-54-9 Molecular Formula: Na3O4P Formula Weight: 163.94 Physical Form: -100 Mesh Powder Sensitivity: Hygroscopic Appearance: White Merck Reference: 14,8662 UN No.: 3262 MDL No.: MFCD00003510
Formel: Na₃O₄P
MW: 163.94 g/mol
Smältpunkt: 73.3…76.7 °C
Densitet: 2.54 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003510
CAS nummer: 7601-54-9
EINECS: 231-509-8
UN: 3262
ADR: 8 III
Merck Index: 14,8662