Sodium hexafluorophosphate ≥99%
Leverantör: Thermo Fisher Scientific
Fara
13064.14 13064.22
13064.14EA 2600 SEK
13064.14 13064.22
Sodium hexafluorophosphate ≥99%
Sodium hexafluorophosphate
Purity: 99+% CAS No.: 21324-39-0 Molecular Formula: NaPF6 Formula Weight: 167.95 Physical Form: Crystalline Density: 2.37 Melting Point: 200° UN No.: 3260 MDL No.: MFCD00011122
Formel: F₆NaP
MW: 167.95 g/mol
Smältpunkt: >200 °C
Densitet: 2.37
Storage Temperature: Omgivande
MDL Number: MFCD00011122
CAS nummer: 21324-39-0
EINECS: 244-333-1
UN: 3260
ADR: 8 II