Litiumhydroxid, vattenfri ≥99.995%
Leverantör: Thermo Scientific
Fara
44471.06 44471.14 44471.22
44471.06EA 1325 SEK
44471.06 44471.14 44471.22
Litiumhydroxid, vattenfri ≥99.995%
Litiumhydroxid
Purity: 99.995% CAS No.: 1310-65-2 Molecular Formula: LiOH Formula Weight: 23.95 Physical Form: Powder Density: 2.54 Appearance: White Odour: Odourless pH: 14 Melting Point: 462° Boiling Point: 924° Merck Reference: 14,5534 UN No.: 2680 MDL No.: MFCD00011095
Formel: LiOH
MW: 23,95 g/mol
Kokpunkt: 924 °C
Smältpunkt: 462 °C
Densitet: 2,54
Storage Temperature: Omgivande
MDL Number: MFCD00011095
CAS nummer: 1310-65-2
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 14,5534