Litiumnitrat, vattenfri ≥99.999%
Leverantör: Thermo Scientific
Fara
Packaged under argon.
44456.09 44456.18 44456.30
44456.09EA 2250 SEK
44456.09 44456.18 44456.30
Litiumnitrat, vattenfri ≥99.999%
Litiumnitrat
Purity: 99.999% CAS No.: 7790-69-4 Molecular Formula: LiNO3 Formula Weight: 68.95 Physical Form: Crystalline/Lump, Packaged under argon Density: 2.38 Appearance: White Odour: Odourless Melting Point: 264° Boiling Point: 600° dec. Merck Reference: 14,5536 UN No.: 2722 MDL No.: MFCD00011094
Formel: LiNO₃
MW: 68,95 g/mol
Kokpunkt: 600 °C (dec.)
Smältpunkt: 264 °C
Densitet: 2,38
Storage Temperature: Omgivande
MDL Number: MFCD00011094
CAS nummer: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1,III
Merck Index: 14,5536