Kaliumnitrat 99.999%
Leverantör: Thermo Scientific
Varning
Synonyms: Nitrate of Potash
193800500 193802500 193800100
ACRO193800500EA 2800 SEK
ACRO193800500 ACRO193802500 ACRO193800100
Kaliumnitrat 99.999%
Potassium nitrate
Formel: KNO₃
MW: 101,1 g/mol
Smältpunkt: 334 °C
Densitet: 2,109 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011409
CAS nummer: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733
Order Now
Specifikation testresultat
Appearance White Crystals and/or chunks
Total trace metal impurities ≤0.001 %