Kaliumpermanganat 99+% ACS
Leverantör: Thermo Fisher Scientific
Fara
424170025 424175000
ACRO424170025EA 3850 SEK
ACRO424170025 ACRO424175000
Kaliumpermanganat 99+% ACS
Kaliumpermanganat
Formel: KMnO₄
Smältpunkt: 240 °C (dec.)
Densitet: 2,7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011364
CAS nummer: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824
Order Now
Specifikation testresultat
Appearance Dark green-black Glistening crystalline powder
Titration with KMnO4 ≥99.0 %
Chloride and Chlorate (as Cl) ≤0.005 %
Sulfate (SO4) ≤0.02 %
Insoluble matter ≤0.2 %