Natriumperjodat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Leverantör: Merck
Fara
Synonyms: Sodium metaperiodate
M-Clarity™ quality level = MQ300
1.06597.0050 1.06597.0250 1.06597.1000
1.06597.0050EA 1570 SEK
1.06597.0050 1.06597.0250 1.06597.1000
Natriumperjodat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Natriumperjodat
Formel: NaIO₄
MW: 213,89 g/mol
Kokpunkt: 300 °C (1013 hPa)
Smältpunkt: 175 °C
Densitet: 3,87 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003534
CAS nummer: 7790-28-5
EINECS: 232-197-6
UN: 3085
ADR: 5.1 (8),I
Merck Index: 13,08714
Order Now
Specifikation testresultat
Solubility 91 g/l (20 °C)
Melting Point 175 °C decomposes
Molar Mass 213.89 g/mol
Bulk Density 2000 - 2400 kg/m3
Boiling Point 300 °C
Density 3.87 g/cm3 (20 °C)
pH 5.2 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer