Natriumperjodat 99.8-100.3% ACS
Leverantör: Thermo Scientific
Fara
Synonyms: Sodium metaperiodate
44309.22 44309.36 44309.A3
44309.22EA 2950 SEK
44309.22 44309.36 44309.A3
Natriumperjodat 99.8-100.3% ACS
Natriumperjodat
Purity: 99.8-100.3% CAS No.: 7790-28-5 Molecular Formula: INaO4 Formula Weight: 213.89 Physical Form: Powder Density: 3.86 Appearance: White Odour: Odourless Melting Point: 300° dec. Merck Reference: 14,8640 UN No.: 1479 MDL No.: MFCD00003534
Formel: NaIO₄
MW: 213,89 g/mol
Kokpunkt: 300 °C (1013 hPa)
Smältpunkt: 300 °C (dec.)
Densitet: 3,86
Storage Temperature: Omgivande
MDL Number: MFCD00003534
CAS nummer: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1,II
Merck Index: 14,8640