Natriumjodid, Wasser_1 99+% Torrvikt
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
A15480.0E A15480.22 A15480.36
A15480.0EEA 14275 SEK
A15480.0E A15480.22 A15480.36
Natriumjodid, Wasser_1 99+% Torrvikt
Natriumjodid
Purity: 99+% CAS No.: 7681-82-5 Molecular Formula: NaI Formula Weight: 149.89 Refractive Index: 1.7745 Odour: Odourless Boiling Point: 1300-1304° Merck Reference: 14,8631 MDL No.: MFCD00003532
Formel: NaI
MW: 149.89 g/mol
Kokpunkt: 1300…1304 °C
Smältpunkt: 651 °C
Densitet: 3.67
Storage Temperature: Omgivande
MDL Number: MFCD00003532
CAS nummer: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9 III
Merck Index: 14,8631