Litiumjodide, vattenfri 99%, ren
Leverantör: Thermo Fisher Scientific
Varning
203590500 203592500
ACRO203590500EA 3425 SEK
ACRO203590500 ACRO203592500
Litiumjodide, vattenfri 99%, ren
Litiumjodide
Formel: ILi
MW: 133.85 g/mol
Kokpunkt: 1177
Smältpunkt: 446 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011092
CAS nummer: 10377-51-2
EINECS: 233-822-5
Merck Index: 13,05557
Order Now
Specifikation testresultat
Appearance White to yellow to beige Powder or crystals or granules
Titration Argentometric ≥98.5 % (on anhydrous substance)
Water ≤1 % (K.F.)