Natriumfluorid ≥99.995%, Puratronic® (K ∼ 60 ppm)
Leverantör: Thermo Fisher Scientific
Fara
12964.06 12964.18 12964.30
12964.06EA 1875 SEK
12964.06 12964.18 12964.30
Natriumfluorid ≥99.995%, Puratronic® (K ∼ 60 ppm)
Natriumfluorid
Purity: 99.995% CAS No.: 7681-49-4 Molecular Formula: NaF Formula Weight: 41.99 Physical Form: Powder Density: 2.78 Refractive Index: 1.336 Odour: Odourless Melting Point: 993° Boiling Point: 1704° Merck Reference: 14,8618 UN No.: 1690 MDL No.: MFCD00003524
Formel: NaF
MW: 41.99 g/mol
Kokpunkt: 1704 °C
Smältpunkt: 993
Densitet: 2.78
Storage Temperature: Omgivande
MDL Number: MFCD00003524
CAS nummer: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1 III
Merck Index: 14,8618
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM