Litiumfluorid ≥99.5%, Optical Grade Optical Grade
Leverantör: Thermo Fisher Scientific
Fara
14056.06 14056.22
14056.06EA 1625 SEK
14056.06 14056.22
Litiumfluorid ≥99.5%, Optical Grade Optical Grade
Litiumfluorid
Purity: 99.5+% CAS No.: 7789-24-4 Molecular Formula: LiF Formula Weight: 25.94 Physical Form: Broken Crystals Density: 2.635 Refractive Index: 1.3915 Odour: Odourless Melting Point: 845° Boiling Point: 1680° Merck Reference: 14,5531 UN No.: 3288 MDL No.: MFCD00011090
Formel: FLi
MW: 25.94 g/mol
Kokpunkt: 1680 °C
Smältpunkt: 845 °C
Densitet: 2.635
Storage Temperature: Omgivande
MDL Number: MFCD00011090
CAS nummer: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1 III
Merck Index: 14,5531