Potassium fluoride dihydrate 98%, ren
Leverantör: Thermo Scientific
Fara
206400050 206402500 206400010
ACRO206400050EA 330 SEK
ACRO206400050 ACRO206402500 ACRO206400010
Potassium fluoride dihydrate 98%, ren
Potassium fluoride dihydrate
Formel: FK·2H₂O
MW: 94,13 g/mol
Kokpunkt: 1505 °C (1013 hPa)
Smältpunkt: 41 °C
Densitet: 2,48 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00149929
CAS nummer: 13455-21-5
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 13,07714
Order Now
Specifikation testresultat
Appearance Colorless to white Crystals and/or chunks
Assay 19.65 to 20.70 % (F)