Natriumklorat ≥99.0% ACS
Leverantör: Thermo Scientific
Fara
44408.22 44408.36 44408.A3
44408.22EA 1050 SEK
44408.22 44408.36 44408.A3
Natriumklorat ≥99.0% ACS
Natriumklorat
Purity: 99.0% min CAS No.: 7775-09-9 Molecular Formula: NaClO3 Formula Weight: 106.45 Physical Form: Crystalline Density: 2.5 Melting Point: 248° Merck Reference: 14,8598 UN No.: 1495 MDL No.: MFCD00003479
Formel: ClNaO₃
MW: 106,44 g/mol
Kokpunkt: 300 °C (1013 hPa)
Smältpunkt: 248 °C
Densitet: 2,5
Storage Temperature: Omgivande
MDL Number: MFCD00003479
CAS nummer: 7775-09-9
EINECS: 231-887-4
UN: 1495
ADR: 5.1,II
REACH: 01-2119474389-23
Merck Index: 14,8598