Kaliumklorat ≥99%
Leverantör: Thermo Fisher Scientific
Fara
A17075.0B A17075.0I A17075.30
A17075.0BEA 1825 SEK
A17075.0B A17075.0I A17075.30
Kaliumklorat ≥99%
Kaliumklorat
Purity: 99+% CAS No.: 3811-04-9 Molecular Formula: KClO3 Formula Weight: 122.55 Odour: Odourless Melting Point: 368° Boiling Point: 400° dec. Merck Reference: 14,7620 UN No.: 1485 MDL No.: MFCD00011361
Formel: KClO₃
Kokpunkt: 400 °C (dec.)
Smältpunkt: 368 °C
Densitet: 2,32
Storage Temperature: Omgivande
MDL Number: MFCD00011361
CAS nummer: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1,II
Merck Index: 14,7620