Natriumklorid ≥99.99%
Leverantör: Thermo Scientific
Varning
|
Synonyms:
Saline, NaCl, Sea salt, Halite
87605.18
87605.30
87605.A1
87605.18EA
1725
SEK
87605.18
87605.30
87605.A1
Natriumklorid ≥99.99%
Natriumklorid
Purity: 99.99%
CAS No.: 7647-14-5
Molecular Formula: NaCl
Formula Weight: 58.44
Physical Form: Crystalline
Density: 2.165
Refractive Index: 1.5442
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
pH: 6-9
Melting Point: 801°
Boiling Point: 1413°
Merck Reference: 14,8599
MDL No.: MFCD00003477
Formel:
NaCl MW: 58,44 g/mol Kokpunkt: 1413 °C Smältpunkt: 801 °C Densitet: 2,165 Storage Temperature: Omgivande |
MDL Number:
MFCD00003477 CAS nummer: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,8599 |
Specifikation testresultat
Total Metal Impurities | 0.01% max |