Litiumklorid, ultra dry ≥99.995%
Leverantör: Thermo Scientific
Varning
Ampuled under argon
13684.06 13684.14 13684.22
13684.06EA 2025 SEK
13684.06 13684.14 13684.22
Litiumklorid, ultra dry ≥99.995%
Litiumklorid
Purity: 99.995% CAS No.: 7447-41-8 Molecular Formula: LiCl Formula Weight: 42.39 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.068 Melting Point: 605° Boiling Point: 1325-1360° Merck Reference: 14,5528 MDL No.: MFCD00011078
Formel: LiCl
MW: 42,39 g/mol
Kokpunkt: 1325…1360 °C
Smältpunkt: 605 °C
Densitet: 2,068
Storage Temperature: Omgivande
MDL Number: MFCD00011078
CAS nummer: 7447-41-8
EINECS: 231-212-3
Merck Index: 14,5528