Natriumbromid ≥97%
Leverantör: Thermo Fisher Scientific
Varning
|
14037.36
14037.A3
14037.36EA
1025
SEK
14037.36
14037.A3
Natriumbromid ≥97%
Sodium bromide
Purity: 97%
CAS No.: 7647-15-6
Molecular Formula: NaBr
Formula Weight: 102.89
Physical Form: Granular
Density: 3.203
Refractive Index: 1.6412
Sensitivity: Hygroscopic
Melting Point: 755°
Boiling Point: 1390°
Solubility: Soluble in water, alcohol
Merck Reference: 14,8594
MDL No.: MFCD00003475
Formel:
NaBr Kokpunkt: 1390 °C Smältpunkt: 755 °C Densitet: 3,203 Storage Temperature: Omgivande |
MDL Number:
MFCD00003475 CAS nummer: 7647-15-6 EINECS: 231-599-9 Merck Index: 14,8594 |