Natriumbromid, vattenfri 99+%, extra pure
Leverantör: Thermo Fisher Scientific
205130010 205131000 205130050
ACRO205130010EA 1375 SEK
ACRO205130010 ACRO205131000 ACRO205130050
Natriumbromid, vattenfri 99+%, extra pure
Sodium bromide
Formel: NaBr
Kokpunkt: 1390 °C (760 mmHg)
Smältpunkt: 755 °C
Densitet: 3,2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003475
CAS nummer: 7647-15-6
EINECS: 231-599-9
Merck Index: 13,08666
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Titration Argentometric ≥99.0 % (on dry substance)
Loss on drying ≤1.0 %
Heavy metals (as Pb) ≤10 ppm