Litiumbromid, vattenfri ≥99.995%
Leverantör: Thermo Scientific
Varning
40657.14 40657.22
40657.14EA 3825 SEK
40657.14 40657.22
Litiumbromid, vattenfri ≥99.995%
Litiumbromid
Purity: 99.995% CAS No.: 7550-35-8 Molecular Formula: BrLi Formula Weight: 86.85 Physical Form: Powder Density: 3.464 Appearance: White Odour: Odourless Melting Point: 550° Boiling Point: 1265° Merck Reference: 14,5526 MDL No.: MFCD00011077
Formel: LiBr
MW: 86,84 g/mol
Kokpunkt: 1265 °C
Smältpunkt: 550 °C
Densitet: 3,464
Storage Temperature: Omgivande
MDL Number: MFCD00011077
CAS nummer: 7550-35-8
EINECS: 231-439-8
Merck Index: 14,5526