Kaliumbromid ≥99%
Leverantör: Thermo Scientific
Varning
A16339.0B A16339.0I A16339.22 A16339.30 A16339.36 A16339.A4
A16339.0BEA 1725 SEK
A16339.0B A16339.0I A16339.22 A16339.30 A16339.36 A16339.A4
Kaliumbromid ≥99%
Potassium bromide
Purity: 99+% CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Refractive Index: 1.559 Odour: Odourless Boiling Point: 1435° Merck Reference: 14,7618 MDL No.: MFCD00011358
Formel: KBr
MW: 119 g/mol
Kokpunkt: 1435 °C
Smältpunkt: 730 °C
Densitet: 2,75
Storage Temperature: Omgivande
MDL Number: MFCD00011358
CAS nummer: 7758-02-3
EINECS: 231-830-3
Merck Index: 14,7618