Natriumtetrafluoroborat 97+%, extra pure
Leverantör: Thermo Scientific
Fara
Synonyms: Sodium fluoroborate
192120250 192125000 192120025
ACRO192120250EA 800 SEK
ACRO192120250 ACRO192125000 ACRO192120025
Natriumtetrafluoroborat 97+%, extra pure
Sodium tetrafluoroborate
Formel: BF₄Na
MW: 109,79 g/mol
Smältpunkt: 384 °C
Densitet: ~ 2,47 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003515
CAS nummer: 13755-29-8
EINECS: 237-340-6
UN: 1759
ADR: 8,II
Merck Index: 12,08761
Order Now
Specifikation testresultat
Appearance White to light grey Crystalline powder or crystals
Titration ≥97.0 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤1000 ppm
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤30 ppm