2-Bromofluorene 95%
Leverantör: Thermo Fisher Scientific
Varning
154220250 154221000
ACRO154220250EA 1825 SEK
ACRO154220250 ACRO154221000
2-Bromofluorene 95%
2-Bromofluorene
Formel: C₁₃H₉Br
MW: 245.12 g/mol
Kokpunkt: 185 °C (135 mmHg)
Smältpunkt: 110…115 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001115
CAS nummer: 1133-80-8
EINECS: 214-480-6
Order Now
Specifikation testresultat
Appearance White to light yellow Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 110 to 115 °C
GC ≥94 %
NMR (1H-NMR) Consistent with structure