1,5-Diazobicyclo[4.3.0]-5-nonene 98
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: DBN, 2,3,4,6,7,8-Hexahydropyrrolo[1,2-a]pyrimidine, 1,5-Diazabicyclo[4.3.0]-5-nonene
A13437.06 A13437.14 A13437.22
A13437.06EA 575 SEK
A13437.06 A13437.14 A13437.22
1,5-Diazobicyclo[4.3.0]-5-nonene 98
1,5-Diazobicyclo[4.3.0]-5-nonene
Purity: 98% CAS No.: 3001-72-7 Molecular Formula: C7H12N2 Formula Weight: 124.19 Flash Point: 94°(201°F) Refractive Index: 1.5200 Boiling Point: 95-98°/7mm Beilstein Reference: 2417 UN No.: 3267 MDL No.: MFCD00005554
Formel: C₇H₁₂N₂
MW: 124.19 g/mol
Kokpunkt: 95…98 °C (7 mmHg)
Densitet: 1.042
Flampunkt: 94 °C (201 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005554
CAS nummer: 3001-72-7
EINECS: 221-087-3
UN: 3267
ADR: 8 II