cis-Cycloocten 95% stabilised
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (Z)-Cyclooctene
Stabilization: Stabilised with IRGANOX 1076
A13477.AD A13477.AE A13477.AK A13477.AP
A13477.ADEA 830 SEK
A13477.AD A13477.AE A13477.AK A13477.AP
cis-Cycloocten 95% stabilised
cis-Cyclooctene
Stabilized
Purity: 95% CAS No.: 931-87-3 Molecular Formula: C8H14 Formula Weight: 110.20 Flash Point: 25°(77°F) Refractive Index: 1.4700 Odour: Recognizable Melting Point: -16° Boiling Point: 145-146° Beilstein Reference: 1280166 UN No.: 3295 MDL No.: MFCD00001753
Formel: C₈H₁₄
Kokpunkt: 145…146 °C
Smältpunkt: –16 °C
Densitet: 0,848
Flampunkt: 25 °C (77 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001753
CAS nummer: 931-87-3
EINECS: 213-243-4
UN: 3295
ADR: 3,III