Bicyclohexyl 99%

Leverantör: Thermo Fisher Scientific

402211000 402210250
ACRO402211000EA 3375 SEK
ACRO402211000 ACRO402210250
Bicyclohexyl 99%
Bicyclohexyl
Formel: C₁₂H₂₂
Kokpunkt: 227 °C
Smältpunkt: 3…4 °C
Densitet: 0,86 g/cm³
Flampunkt: 92 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003815
CAS nummer: 92-51-3
EINECS: 202-161-4

Order Now

Specifikation testresultat

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4800 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR