Ethylcyclohexane 99+%
Leverantör: Thermo Scientific
Fara
118310250 118311000
ACRO118310250EA 870 SEK
ACRO118310250 ACRO118311000
Ethylcyclohexane 99+%
Ethylcyclohexane
Formel: C₈H₁₆
MW: 112,22 g/mol
Kokpunkt: 132 °C (1013 hPa)
Smältpunkt: –111 °C
Densitet: 0,78 g/cm³
Flampunkt: 18 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001523
CAS nummer: 1678-91-7
EINECS: 216-835-0
UN: 3295
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4311 to 1.4331 (20°C, 589 nm)
GC ≥99.0 %
Clarity of solution Passes test