4-tert-Butylcyclohexylamine cis- and trans-mixture 97%
Leverantör: Thermo Fisher Scientific
Fara
L02472.06 L02472.14
L02472.06EA 1050 SEK
L02472.06 L02472.14
4-tert-Butylcyclohexylamine cis- and trans-mixture 97%
4-tert-Butylcyclohexylamine
Purity: 97% CAS No.: 5400-88-4 Molecular Formula: C10H21N Formula Weight: 155.29 Flash Point: 79°(174°F) Refractive Index: 1.4662 Beilstein Reference: 1523780 UN No.: 2735 MDL No.: MFCD00013763
Formel: C₁₀H₂₁N
Kokpunkt: 202 °C (1013 hPa)
Smältpunkt: 18 °C
Densitet: 0,870
Flampunkt: 79 °C (174 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00013763
CAS nummer: 5400-88-4
EINECS: 226-444-7
UN: 2735
ADR: 8,III