1-Methylcyclohexanecarboxylic acid ≥99%
Leverantör: Thermo Scientific
Synonyms: 1-Methyl-1-cyclohexanecarboxylic acid, 1-Methylcyclohexane-1-carboxylic acid
B21713.06 B21713.14 B21713.18
B21713.06EA 1125 SEK
B21713.06 B21713.14 B21713.18
1-Methylcyclohexanecarboxylic acid ≥99%
1-Methylcyclohexanecarboxylic acid
Purity: 99% CAS No.: 1123-25-7 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 100°(212°F) Melting Point: 36-39° Boiling Point: 236° Beilstein Reference: 1859220 MDL No.: MFCD00001463
Formel: C₈H₁₄O₂
MW: 142,2 g/mol
Kokpunkt: 236 °C
Smältpunkt: 36…39 °C
Densitet: 1,0115 g/cm³ (20 °C)
Flampunkt: 100 °C (212 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001463
CAS nummer: 1123-25-7
EINECS: 214-371-3