5-Methyl-1,3-cyclohexanedione ≥98%
Leverantör: Thermo Fisher Scientific
|
Varning
|
Synonyms:
5-Methylcyclohexane-1,3-dione
A15280.03
A15280.06
A15280.14
A15280.03EA
1075
SEK
A15280.03
A15280.06
A15280.14
5-Methyl-1,3-cyclohexanedione ≥98%
5-Methyl-1,3-cyclohexanedione
Purity: 98%
CAS No.: 4341-24-6
Molecular Formula: C7H10O2
Formula Weight: 126.16
Melting Point: 126-130°
Beilstein Reference: 1099692
Formel:
C₇H₁₀O₂ Smältpunkt: 126…130 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00010379 CAS nummer: 4341-24-6 |