2-Methyl-1,3-cyclopentanedione ≥98%
Leverantör: Thermo Scientific
Synonyms: 2-Methylcyclopentane-1,3-dione
A13936.06 A13936.14 A13936.22
A13936.06EA 1200 SEK
A13936.06 A13936.14 A13936.22
2-Methyl-1,3-cyclopentanedione ≥98%
2-Methyl-1,3-cyclopentanedione
Purity: 98% CAS No.: 765-69-5 Molecular Formula: C6H8O2 Formula Weight: 112.13 Melting Point: 212-216° Beilstein Reference: 1237255
Formel: C₆H₈O₂
MW: 112,13 g/mol
Kokpunkt: 213 °C (1013 hPa)
Smältpunkt: 212…216 °C
Densitet: 1,33 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00001406
CAS nummer: 765-69-5
EINECS: 212-153-2