2-Methyl-1,3-cyclohexanedione ≥98%
Leverantör: Thermo Fisher Scientific
Synonyms: 2-Methylcyclohexane-1,3-dione
A11218.06 A11218.14 A11218.22
A11218.06EA 815 SEK
A11218.06 A11218.14 A11218.22
2-Methyl-1,3-cyclohexanedione ≥98%
2-Methyl-1,3-cyclohexanedione
Purity: 98+% CAS No.: 1193-55-1 Molecular Formula: C7H10O2 Formula Weight: 126.16 Storage Temperature: 2°C to 8°C Melting Point: 204-208° Beilstein Reference: 1281598 MDL No.: MFCD00001587
Formel: C₇H₁₀O₂
Kokpunkt: 100…102 °C (8 mmHg)
Smältpunkt: 204…208 °C
Densitet: 1,313 g/cm³ (–150.15 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00001587
CAS nummer: 1193-55-1
EINECS: 214-773-9