2-Methylcyclopentanone 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Methylcyclopentan-1-one
247660050 247660250
ACRO247660050EA 1950 SEK
ACRO247660050 ACRO247660250
2-Methylcyclopentanone 99%
2-Methylcyclopentanone
Formel: C₆H₁₀O
MW: 98,14 g/mol
Kokpunkt: 139…140 °C
Smältpunkt: -75 °C
Densitet: 0,91 g/cm³
Flampunkt: 26 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001414
CAS nummer: 1120-72-5
EINECS: 214-318-4
UN: 1224
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20°C, 589 nm)
GC ≥98.5 %