(-)-Menthone ≥90.0%

Leverantör: TCI EUROPE

Synonyms: trans-Menthone, L-Menthone, (2S,5R)-2-Isopropyl-5-methylcyclohexanone, (1R,4S)-p-Menthan-3-one

M0513-25ML M0513-500ML M0513-100ML
TCIAM0513-25MLEA 295 SEK
TCIAM0513-25ML TCIAM0513-500ML TCIAM0513-100ML
(-)-Menthone ≥90.0%
(-)-Menthone
Formel: C₁₀H₁₈O
MW: 154,25 g/mol
Kokpunkt: 88°C
Flampunkt: 72°C
MDL Number: MFCD00001634
CAS nummer: 14073-97-3
EINECS: 237-926-1
Merck Index: 14,05838
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2aff82f8028b19af3f2f3c36df425ee07680e4e1&vid=2aff82f8028b19af3f2f3c36df425ee07680e4e1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %
Specific rotation [a]20/D -15.0 to -24.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR