Saltsyra 2 mol/l (2 N), SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Varning
Synonyms: Hydrochloric acid solution, Muriatic acid
4.80934.1000 4.80934.9025
4.80934.1000EA 2000 SEK
4.80934.1000 4.80934.9025
Saltsyra 2 mol/l (2 N), SAFC®
Saltsyra
Formel: HCl
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II