TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Millipore®
Leverantör: Merck
Varning
Synonyms: TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid
M-Clarity™ quality level = MQ100
1.10695.0250 1.10695.9010
1.10695.0250EA 4775 SEK
1.10695.0250 1.10695.9010
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Millipore®
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Formel: C₆H₁₅NO₆S
Storage Temperature: Omgivande
MDL Number: MFCD00007532
CAS nummer: 7365-44-8
EINECS: 230-906-3